Environmental Express SimpleDist® Washers for 38 mm Cap; 6/Pk

Representative image only.

Environmental Express SimpleDist® Washers for 38 mm Cap; 6/Pk

Environmental Express | Item: 3520993

Individual components for your SimpleDist system

₹3,000.00INR / Pkg of 6

₹3,000.00INR / Pkg of 6
Product Description

Key Features

  • Replacement manifolds and washers
  • Tubing and adapters can be replaced after wear and tear
  • Collection traps for safe pouring of the final solution
  • 3 month warranty

More About this Item

Ensure cyanide and ammonia collection run smoothly by replacing parts when appropriate.

Washers for 38mm open-top screw caps 35209-92 (C6120) provide a tight, leak-resistant seal.

3 Month Warranty
Specifications
  • Compatible With38 mm open-top screw caps (C6120)
  • Diameter (mm)38
  • DescriptionSimpleDist Washers for 38 mm Cap; 6/Pk
  • Warranty3 month
Reviews
Q&A