Environmental Express SimpleDist® Two-Port Cap Inserts; 6/Pk
Representative image only.
Environmental Express SimpleDist® Two-Port Cap Inserts; 6/Pk
Environmental Express | Item: 3520994
Individual components for your SimpleDist system
₹5,900.00INR / Pkg of 6
₹5,900.00INR / Pkg of 6
Product Description
Key Features
  • Replacement manifolds and washers
  • Tubing and adapters can be replaced after wear and tear
  • Collection traps for safe pouring of the final solution
  • 3 month warranty
More About this Item
Ensure cyanide and ammonia collection run smoothly by replacing parts when appropriate.
The unique two-port cap insert fits snugly into the open-top screw cap. The cap insert features a luer taper design to connect the collection trap to the boiling flask.
Disposable acid addition tubes with funnel tops slide easily, but firmly, into the bo
3 Month Warranty
Specifications
  • MaterialPolypropylene (PP)
  • Compatible WithOpen-top screw cap (C6120)
  • Diameter (mm)38
  • DescriptionSimpleDist Two-Port Cap Inserts; 6/Pk