Environmental Express SimpleDist® Tubing Kit; 12 Pieces/Pk
Representative image only.
Environmental Express SimpleDist® Tubing Kit; 12 Pieces/Pk
Environmental Express | Item: 3520996
Individual components for your SimpleDist system
₹19,500.00INR / Pkg of 12
₹19,500.00INR / Pkg of 12
Product Description
Key Features
  • Replacement manifolds and washers
  • Tubing and adapters can be replaced after wear and tear
  • Collection traps for safe pouring of the final solution
  • 3 month warranty
More About this Item
Ensure cyanide and ammonia collection run smoothly by replacing parts when appropriate.
Tubing kits come with 12 lengths of tubing. Six pieces of tubing are 8" and the other 6 are 10.25". The longer pieces extend from the manifold valves to the front row of sample collection flasks while the shorter lengths go to the back row of collection f
3 Month Warranty
Specifications
  • MaterialPolypropylene (PP)
  • Compatible WithSimpleDist System
  • DescriptionSimpleDist Tubing Kit; 12 Pieces/Pk