Environmental Express SimpleDist Tubing Adapters; 12/Pk
Representative image only.
Environmental Express SimpleDist Tubing Adapters; 12/Pk
Environmental Express | Item: 3520995
Individual components for your SimpleDist system
₹7,000.00INR / Pkg of 12
₹7,000.00INR / Pkg of 12
Product Description
Key Features
  • Replacement manifolds and washers
  • Tubing and adapters can be replaced after wear and tear
  • Collection traps for safe pouring of the final solution
  • 3 month warranty
More About this Item
Ensure cyanide and ammonia collection run smoothly by replacing parts when appropriate.
The narrow end of the tubing adaptor fits tightly into 1/4" ID tubing while the larger open end fits securely onto the top of the cyanide trap.
3 Month Warranty
Specifications
  • MaterialPolypropylene (PP)
  • Compatible WithSimpleDist cyanide trap
  • DescriptionSimpleDist Tubing Adapters; 12/Pk