Environmental Express SimpleDist® Complete Distillation System; 120 VAC

Representative image only.

Environmental Express SimpleDist® Complete Distillation System; 120 VAC

Environmental Express | Item: 3520984

Safe and easy distillation system – the name says it all!

Call For Price

Call For Price

Buy It Together

Current Item

Environmental Express SimpleDist® Complete Distillation System; 120 VAC
Environmental Express SimpleDist® Complete Distillation System; 120 VAC
3520984

Call For Price
+
Environmental Express SimpleDist® Collection Traps, 25 mL; 100/Pk
3520990

₹59,000.00INR / Pkg of 100
=
Total Price 59000.00 ADD 2 ITEMS TO CART
Product Description

Includes

12-place HotBlock, 12-valve manifold, 12 boiling tubes, 1 pk 25-mL collection traps, 1 pk of reagent addition tubes, caps, washers, inserts, and tubing kit.

Key Features

  • Conform to EPA and Standard Methods
  • System reduces labor and clean-up time
  • Unique design replaces much of the fragile glassware usually involved in such procedures
  • 1 year warranty

More About this Item

The optimal system for performing cyanide and ammonia distillations following established EPA methods 335.4 and 350.1, and Standard Methods 4500-CN-C and 4500-NH3.

The distillate is collected in a disposable trap, and does not require the use of a chiller or water lines. The sample boils in the glass boiling tube, and the gas is pulled by vacuum into the trap where it is collected. A hydrophobic porous frit in the trap prevents solution from flowing back into the boiling tube. When complete, the trap is removed and the contents are ready for analysis — Simple!

1 Year Warranty
Specifications
  • Tube Size (mL)50
  • Set-Point Accuracy±0.2°F (0.1°C)
  • Max Operating Temperature (° F)302
  • Max Operating Temperature (° C)150
  • Height (in)25
  • Width (in)19 1/2
  • Depth (in)11 1/4
  • Height (cm)63.5
  • Width (cm)49.5
  • Depth (cm)28.6
  • Power (VAC)120
  • Power (Hz)60
  • Power (amps)6
  • DescriptionComplete Distillation System; 120 VAC
  • Warranty1 year
Reviews
Q&A