Environmental Express SimpleDist® Acid Addition Tube with Funnel Tip; 100/Pk
Representative image only.
Environmental Express SimpleDist® Acid Addition Tube with Funnel Tip; 100/Pk
Environmental Express | Item: 3520991
Individual components for your SimpleDist system
₹7,100.00INR / Pkg of 100
₹7,100.00INR / Pkg of 100
Product Description
Key Features
  • Replacement manifolds and washers
  • Tubing and adapters can be replaced after wear and tear
  • Collection traps for safe pouring of the final solution
  • 3 month warranty
More About this Item
Ensure cyanide and ammonia collection run smoothly by replacing parts when appropriate.
Disposable polypropylene acid addition and inlet tubes with funnel tips replace fragile glass tubing in traditional systems. The tubes slide easily but firmly into the boiling flask through the two-port cap insert.
3 Month Warranty
Specifications
  • MaterialPolypropylene (PP)
  • Compatible WithSimpleDist System
  • DescriptionSimpleDist Acid Addition Tube with Funnel Tip; 100/Pk