Environmental Express SimpleDist 12-Place Manifold Kit without Block
Representative image only.
Environmental Express SimpleDist 12-Place Manifold Kit without Block
Environmental Express | Item: 3520987
Individual components for your SimpleDist system
₹777,000.00INR / Each
₹777,000.00INR / Each
Product Description
Includes
12-valve stainless steel manifold, 12 flow control valves, 1 barb connector, 12 boiling tubes, 1 pack of (C6100) 25 mL collection traps, 1 pack of (C6110) reagent addition tubes, 2 packs of (C6120) caps, 2 packs of (C6120A) black neoprene washers, 2 packs of (C6130) two-port cap inserts, and 1 (C6142) tubing kit.
Key Features
  • Replacement manifolds and washers
  • Tubing and adapters can be replaced after wear and tear
  • Collection traps for safe pouring of the final solution
  • The SimpleDist System uses the traditional chemistry found in EPA Method 335.4, and Standard Methods 4500-CN-C
  • 1 year warranty
More About this Item
Ensure cyanide and ammonia collection run smoothly by replacing parts when appropriate.
Convenient, disposable cyanide and ammonia collection traps provide a low cost alternative to completely disposable systems while allowing the analyst to use a traditional 50mL sample size. The 12-position system eliminates most of the glassware associated with traditional systems. The system does not use cooling water or the cumbersome tubing that cooling water requires, making it easy to use and inexpensive to maintain and operate.
HotBlock not included.
1 Year Warranty
Specifications
  • MaterialStainless steel
  • Compatible WithSC100 HotBlock
  • DescriptionSimpleDist 12-Place Manifold Kit without Block