Environmental Express pH Electrodes for Field Use; double-junction 10 foot cable
Representative image only.
Environmental Express pH Electrodes for Field Use; double-junction 10 foot cable
Environmental Express | Item: 0599206
Ideal for your most rugged applications
₹30,600.00INR / Each
₹30,600.00INR / Each
Buy It Together
Current Item
Environmental Express pH Electrodes for Field Use; double-junction 10 foot cable
Environmental Express pH Electrodes for Field Use; double-junction 10 foot cable
0599206
₹30,600.00INR / Each
+ + + + =
Total Price 290100.00 ADD 5 ITEMS TO CART
Product Description
Key Features
  • Ceramic reference-junction pH electrodes (clog resistant)
  • 1 year warranty
More About this Item
These combination electrodes withstand the most demanding field conditions—the semi-domed tip protects the pH bulb and ensures a quick response when taking measurements in harsh and dirty environments. Easy-to-clean electrodes feature an impact-resistant epoxy body for further protection from the elements. The large, annular ceramic reference junction and sealed cross-linked polymer reference resist clogging.
Operating range is from 0 to 14 pH. Electrodes measure 110 mm L x 12 mm dia and withstand temperatures up to 212°F (100°C). All models have BNC connectors.
1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Min pH (pH)14
  • Max Temperature (° C)100
  • ConnectorBNC
  • Cable Length (m)3
  • DescriptionpH Electrodes for Field Use; double-junction 10 foot cable