Environmental Express pH electrode, Refillable, long and thin body
Representative image only.
Environmental Express pH electrode, Refillable, long and thin body
Environmental Express | Item: 0599142
Faster response and increased stability across a pH range of 0 to 14
₹39,900.00INR / Each
₹39,900.00INR / Each
Buy It Together
Current Item
Environmental Express pH electrode, Refillable, long and thin body
Environmental Express pH electrode, Refillable, long and thin body
0599142
₹39,900.00INR / Each
+ =
Total Price 51100.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
  • 6 month warranty
More About this Item
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.
6 Month Warranty
Specifications
  • ApplicationsGeneral purpose-Thin neck
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • ConnectorBNC
  • Diameter (mm)8
  • Length (mm)154
  • DescriptionpH electrode, Refillable, long and thin body
  • Warranty6 month warranty