Environmental Express Overcaps for BOD Bottles, PE; 50/Pk
Representative image only.
Environmental Express Overcaps for BOD Bottles, PE; 50/Pk
Environmental Express | Item: 5320014
Using a fresh, clean bottle for each analysis provides more accurate results in your lab
₹12,100.00INR / Pkg of 50
₹12,100.00INR / Pkg of 50
Product Description
Key Features
  • Ideal for 300 mL BOD bottles
  • 3 month warranty
More About this Item
The use of a cap over the mouth of a the BOD bottle to prevent evaporation is recommended by Standard Methods. Our overcaps are made of polyethylene and fit snugly against the rim and shoulder of our disposable BOD bottles. These overcaps do not fit the glass BOD bottles.
BOD overcaps fit tightly over BOD bottles to prevent evaporation. Required by Standard Methods.
3 Month Warranty
Specifications
  • MaterialPolyethylene
  • Compatible With300 mL plastic BOD bottles
  • DescriptionOvercaps for BOD Bottles, PE; 50/Pk