Environmental Express ORP Electrode, Sealed, Waste water, reaction monitoring
Representative image only.
Environmental Express ORP Electrode, Sealed, Waste water, reaction monitoring
Environmental Express | Item: 0599118
Faster response and increased stability across a pH range of 0 to 14
₹38,500.00INR / Each
₹38,500.00INR / Each
Buy It Together
Current Item
Environmental Express ORP Electrode, Sealed, Waste water, reaction monitoring
Environmental Express ORP Electrode, Sealed, Waste water, reaction monitoring
0599118
₹38,500.00INR / Each
+ =
Total Price 49700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
  • 1 year warranty
More About this Item
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.
1 Year Warranty
Specifications
  • ApplicationsDirty Samples, Waste water
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • ConnectorBNC
  • DescriptionORP Electrode, Sealed, Waste water, reaction monitoring