Environmental Express Oakton pH Electrode, Epoxy, Double-Junction, Gel-Filled, 10-ft BNC Cable
Representative image only.
Environmental Express Oakton pH Electrode, Epoxy, Double-Junction, Gel-Filled, 10-ft BNC Cable
Update, upgrade, or replace electrodes to get the best out of Oakton® meters
₹31,000.00INR / Each
₹31,000.00INR / Each
Buy It Together
Current Item
Environmental Express Oakton pH Electrode, Epoxy, Double-Junction, Gel-Filled, 10-ft BNC Cable
Environmental Express Oakton pH Electrode, Epoxy, Double-Junction, Gel-Filled, 10-ft BNC Cable
3580524
₹31,000.00INR / Each
+ + + + +
Cole-Parmer 100-series Replacement pH/Temperature Electrode
5920006
₹22,100.00INR / Each
+
Environmental Express Buffer Solution, pH 12.46; 500 mL
0065412
₹2,100.00INR / Each
+ + + =
Total Price 364100.00 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from pH or ORP electrodes
  • Submersible pH sensor measures 6.00 inches length and 1.05 inch in diameter
  • 1 year warranty
More About this Item
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
1 Year Warranty
Specifications
  • MaterialEpoxy
  • Compatible WithOakton pH/Con 300
  • Length (InchShort)6
  • Diameter (InchShort)1 1/20
  • DescriptionpH Probe, Epoxy Body, Double-Junction, Gel-Filled, 10-ft Cable; BNC