Environmental Express Oakton Ion Selective Electrode (Water hardness Ca/Mg)
Representative image only.
Environmental Express Oakton Ion Selective Electrode (Water hardness Ca/Mg)
Environmental Express | Item: 3581248
A wide variety of ion selective electrodes are availabe to suit a wide array of applications
₹62,300.00INR / Each
₹62,300.00INR / Each
Buy It Together
Current Item
Environmental Express Oakton Ion Selective Electrode (Water hardness Ca/Mg)
Environmental Express Oakton Ion Selective Electrode (Water hardness Ca/Mg)
3581248
₹62,300.00INR / Each
+
Cole-Parmer Copper (Cu+2) ISE Double junction solution kits
2750265
₹22,300.00INR / Each
+
Cole-Parmer Replaceable Membrane ISE Probe; Ammonium
2750500
₹64,600.00INR / Each
+ +
Environmental Express PC 2000 Benchtop Meter Kit
3541965
₹242,000.00INR / Each
+ + + =
Total Price 790200.00 ADD 8 ITEMS TO CART
Product Description
Key Features
  • Epoxy-body types are either single- or double-junction
  • Glass-body type is double junction
  • 1 year warranty
More About this Item
Select from four electrode types: membrane, solid-state, gas sensing, and glass bulb; typical response time is 20 to 30 seconds.
1 Year Warranty
Specifications
  • Electrode TypeDouble junction, membrane
  • Electrode Body MaterialEpoxy
  • Ion-Selective ElectrodeWater hardness Ca/Mg
  • ConnectorBNC
  • DescriptionIon Selective Electrode (Water hardness Ca/Mg)