Environmental Express Oakton CERTIFIED pH CRM Buffer, Red, pH 4; 500 mL

Representative image only.

Environmental Express Oakton CERTIFIED pH CRM Buffer, Red, pH 4; 500 mL

pH buffers tested and certified for laboratory compliance

₹6,300.00INR / Each

₹6,300.00INR / Each

Buy It Together

Current Item

Environmental Express Oakton CERTIFIED pH CRM Buffer, Red, pH 4; 500 mL
Environmental Express Oakton CERTIFIED pH CRM Buffer, Red, pH 4; 500 mL
0065106

₹6,300.00INR / Each
+ + +
Environmental Express Electrode Storage Solution; 500 mL
0065304

₹3,700.00INR / Pint of 1
+ + + =
Total Price 394900.00 ADD 7 ITEMS TO CART
Product Description

Key Features

  • Certified Reference Material for compliance-controlled laboratories and facilities
  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color-coded and/or clearly labeled for easy identification in busy labs
  • Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory
  • NIST/USA buffer value of pH 4.005 traceable to NIST standard reference materials

More About this Item

Certified Reference Materials (CRM) meet federal, state, and local requirements, providing standardized reference calibration for pH meters. Produced in compliance with ISO/IEC 17025:2017, ISO 17034:2016, ISO Guide 31, and ISO 33405, CRMs ensure top-tier accuracy, documentation, and standards. They undergo extensive stability, transportation, and repeatability testing, with uncertainty (U) assigned from these evaluations. Calibration reports are issued by an ISO 17034 and ISO 17025 accredited lab. Environmental Express Oakton monitors the solution over 1 to 2 years and informs users of any changes.

pH 4 is a common high-range pH meter calibration point. All Oakton pH buffer solutions are manufactured under ISO 9000 quality guidelines and traceable to NIST standard (for meters with 0.01 resolution). A pH/temperature table is printed on the label for accurate calibrations. Available as Reference Materials (RM) and Certified Reference Material (CRM) grade. pH 4 buffer solution is available colored red for visual reference or clear. It is sold in individual 500-mL bottles. If unopened, shelf life of pH 4 is 2 years. Store buffers away from direct sunlight.

NIST Traceable Calibration
Specifications
  • Buffer Value4.005
  • Volume (mL)500
  • ColorRed
  • DescriptionEnvironmental Express Oakton CERTIFIED pH CRM Buffer, Red, pH 4; 500 mL
Documents & Certificates

Certificate of Analysis

Enter lot number
or click to view a Sample C of A
Reviews
Q&A