Environmental Express Oakton CERTIFIED CRM pH Buffer, Blue, pH 10; 4 L

Representative image only.

Environmental Express Oakton CERTIFIED CRM pH Buffer, Blue, pH 10; 4 L

pH buffers tested and certified for laboratory compliance

₹19,100.00INR / Each

₹19,100.00INR / Each

Buy It Together

Current Item

Environmental Express Oakton CERTIFIED CRM pH Buffer, Blue, pH 10; 4 L
Environmental Express Oakton CERTIFIED CRM pH Buffer, Blue, pH 10; 4 L
0065180

₹19,100.00INR / Each
+ + + + + +
Environmental Express Electrode Storage Solution; 500 mL
0065304

₹3,700.00INR / Pint of 1
=
Total Price 407700.00 ADD 7 ITEMS TO CART
Product Description

Key Features

  • Certified Reference Material for compliance-controlled laboratories and facilities
  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color-coded and/or clearly labeled for easy identification in busy labs
  • Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory
  • 4 L (128 fl.oz.) capacity
  • Calibrate at pH 10
  • Easily identify blue color-coded formula

More About this Item

Certified Reference Materials (CRM) meet federal, state, and local requirements, providing standardized reference calibration for pH meters. Produced in compliance with ISO/IEC 17025:2017, ISO 17034:2016, ISO Guide 31, and ISO 33405, CRMs ensure top-tier accuracy, documentation, and standards. They undergo extensive stability, transportation, and repeatability testing, with uncertainty (U) assigned from these evaluations. Calibration reports are issued by an ISO 17034 and ISO 17025 accredited lab. Environmental Express Oakton monitors the solution over 1 to 2 years and informs users of any changes.

If unopened, shelf life is 2 years from the date of manufacture; once opened, use within 60 days. Store buffers away from direct sunlight.

NIST Traceable Calibration
Specifications
  • Buffer Value10.012
  • Volume (mL)4000
  • ColorBlue
  • DescriptionEnvironmental Express Oakton CERTIFIED CRM pH Buffer, Blue, pH 10; 4 L
Documents & Certificates

Certificate of Analysis

Enter lot number
or click to view a Sample C of A
Reviews
Q&A