Environmental Express Oakton CERTIFIED CRM One-Shot™ Conductivity and TDS Standard, 1413 µS; 6 x 100 mL Vials

Representative image only.

Environmental Express Oakton CERTIFIED CRM One-Shot™ Conductivity and TDS Standard, 1413 µS; 6 x 100 mL Vials

Environmental Express Oakton conductivity standards are Certified Reference Materials, making one of the most accurate available

₹6,000.00INR / Pkg of 6

₹6,000.00INR / Pkg of 6

Buy It Together

Current Item

Environmental Express Oakton CERTIFIED CRM One-Shot™ Conductivity and TDS Standard, 1413 µS; 6 x 100 mL Vials
Environmental Express Oakton CERTIFIED CRM One-Shot™ Conductivity and TDS Standard, 1413 µS; 6 x 100 mL Vials
0065268

₹6,000.00INR / Pkg of 6
+
Environmental Express Electrode Storage Solution; 500 mL
0065304

₹3,700.00INR / Pint of 1
+
Environmental Express Buffer Solution, pH 12.46; 500 mL
0065412

₹2,100.00INR / Each
+ =
Total Price 16400.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Temperature compensation chart, TDS conversion chart, and NIST-traceable calibration certificate with data.

Key Features

  • All Environmental Express Oakton conductivity standards are NIST-traceable reference materials
  • Accredited to ISO 17034 requirements
  • These standard solutions let you calibrate conductivity instruments to ensure accuracy
  • Conductivity options range from 1 to 200,000 µS/cm
  • If unopened, shelf life is one year; once opened, shelf life will vary by solution value
  • Use when readings are expected to be around 1413 µS/cm

More About this Item

Environmental Express Oakton One-Shot™ conductivity and TDS standards include detailed KCl, NaCl, and 442 conversions, temperature corrections, and certified values referenced to 25°C—convenient. Average accuracy at 25°C is ±0.25 microsiemens for 1, 5, 10 microsiemen (µS) solutions, or ±0.25% for all other solutions. The tested uncertainty is shown on the certificate, whichever is greater.

Batch-tested solutions are lot certified; a few bottles from a single lot are analyzed.

METHODS SUPPORTED
Certified reference material meets test requirements for Federal, State, and local agencies, CAP, ASTM, NCCLS, CLSI, ACS, CLIA, AOAC, EPA, APHA, AWWA, WEF, USGS, USP, and ISO. Traceable® Conductivity Standards complies with and is essential for use in these official methods: AOAC 973.40, EPA 120.1, Standard Method 2510 (APHA, AWWA, WEF), ISO 7888, DIN 38404, ASTM D1125, USGS I-1780, USP 645, and for A2LA/NVLAP accreditations/ISO 9000 certifications. Material may be used to calibrate all conductivity meters and to determine all conductivity cell constants.

A2LA ACCREDITATION ISO/IEC 17025:2005 (1750.01)
To assure accuracy an individually serial-numbered Traceable® Certificate is supplied to indicate traceability to standards provided by NIST (National Institute of Standards and Technology) and/or a National Standards Laboratory. A2LA accredited certification provides the highest achievable level of quality assurance, documentation, and accuracy. Conductivity Standards are produced in an A2LA accredited ISO/IEC 17025:2005 (1750.01) calibration laboratory. Additional certifications include ISO 31 (certificate content) and ISO 35 (statistical analysis), plus ISO 9001 (certified quality manufacturer).

Guaranteed to have at least 90 days remaining on certificate.

NIST Traceable Calibration
Specifications
  • Conductivity Value (µS)1413
  • Conductivity Value (mS)1.413
  • Conductivity Value1413 µS/cm
  • TDS Value (ppm)933
  • Volume (mL)100
  • Volume (oz)3.4
  • Volume6 x 100 mL vials
  • DescriptionCERTIFIED CRM One-Shot™ Conductivity and TDS Standard, 1413 µS; 6 x 100 mL Vials
Documents & Certificates
Reviews
Q&A