Environmental Express Oakton CERTIFIED CRM One-Shot™ Buffer Solution, Clear, pH 2.000
Representative image only.
Environmental Express Oakton CERTIFIED CRM One-Shot™ Buffer Solution, Clear, pH 2.000
Convenient, single-use vials eliminate concerns of cross-contamination
₹7,000.00INR / Each
₹7,000.00INR / Each
Product Description
Includes
Each pack contains six polyethylene vials with 100-mL of pH 2.000 buffer per vial, individual temperature compensation chart, and NIST-Traceable calibration certificate with data.
Key Features
  • Certified Reference Material for compliance-controlled laboratories and facilities
  • Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory
  • Individually packed in foil pouch to prevent UV exposure that may degrade accuracy
  • Buffer solution lets you calibrate pH instruments to ensure accuracy
  • If unopened, shelf life is 2 years
More About this Item
Environmental Express® Oakton® One-Shot™ pH Buffer Solutions are supplied with NIST-traceable calibration reports stating accuracy at 25°C to ±0.010 pH, in addition to conversion values for pH/temperature—for the most accurate calibrations!
Certified Reference Materials (CRM) meet federal, state, and local requirements, providing standardized reference calibration for pH meters. Produced in compliance with ISO/IEC 17025:2017, ISO 17034:2016, ISO Guide 31, and ISO 33405, CRMs ensure top-tier accuracy, documentation, and standards. They undergo extensive stability, transportation, and repeatability testing, with uncertainty (U) assigned from these evaluations. Calibration reports are issued by an ISO 17034 and ISO 17025 accredited lab. Environmental Express® Oakton® monitors the solution over 1 to 2 years and informs users of any changes.
Specifications
  • Buffer Value2
  • Volume (mL)600
  • ColorClear
  • DescriptionCERTIFIED CRM One-Shot™ Buffer Solution, Clear, pH 2.000