Environmental Express Oakton CERTIFIED CRM Conductivity and TDS Standard, Individually-Tested, 1000 µS; 500 mL

Representative image only.

Environmental Express Oakton CERTIFIED CRM Conductivity and TDS Standard, Individually-Tested, 1000 µS; 500 mL

Environmental Express Oakton CERTIFIED conductivity standards are Certified Reference Materials, making one of the most accurate available

₹15,900.00INR / Each

₹15,900.00INR / Each

Buy It Together

Current Item

Environmental Express Oakton CERTIFIED CRM Conductivity and TDS Standard, Individually-Tested, 1000 µS; 500 mL
Environmental Express Oakton CERTIFIED CRM Conductivity and TDS Standard, Individually-Tested, 1000 µS; 500 mL
0065248

₹15,900.00INR / Each
+ +
Environmental Express Buffer Solution, pH 12.46; 500 mL
0065412

₹2,100.00INR / Each
+
Environmental Express Electrode Storage Solution; 500 mL
0065304

₹3,700.00INR / Pint of 1
=
Total Price 27000.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

NIST-traceable calibration certificate of Standard.

Key Features

  • All Environmental Express Oakton CERTIFIED conductivity standards are NIST-traceable reference materials
  • Certified to ISO/IEC 17025 requirements
  • These standard solutions let you calibrate conductivity instruments to ensure accuracy
  • Conductivity options range from 1 to 200,000 µS/cm
  • If unopened, shelf life is one year; once opened, shelf life will vary by solution value
  • Use when readings are expected to be around 1000 µS/cm

More About this Item

Environmental Express Oakton CERTIFIED conductivity and TDS standards have detailed labels: KCl, NaCl, and 442 conversions, temperature corrections, and certified values referenced to 25°C—convenient. Average accuracy at 25°C is ±0.25 microsiemens for 1, 5, 10 microsiemen (µS) solutions, or ±0.25% for all other solutions. The tested uncertainty is shown on the certificate, whichever is greater.

Individually-tested solutions are certified by the bottle—each bottle is analyzed for the best accuracy

METHODS SUPPORTED
Certified reference material meets test requirements for Federal, State, and local agencies, CAP, ASTM, NCCLS, CLSI, ACS, CLIA, AOAC, EPA, APHA, AWWA, WEF, USGS, USP, and ISO. Traceable® Conductivity Standards complies with and is essential for use in these official methods: AOAC 973.40, EPA 120.1, Standard Method 2510 (APHA, AWWA, WEF), ISO 7888, DIN 38404, ASTM D1125, USGS I-1780, USP 645, and for A2LA/NVLAP accreditations/ISO 9000 certifications. Material may be used to calibrate all conductivity meters and to determine all conductivity cell constants.

A2LA ACCREDITATION ISO/IEC 17025:2005 (1750.01)
To assure accuracy an individually serial-numbered Traceable® Certificate is supplied to indicate traceability to standards provided by NIST (National Institute of Standards and Technology) and/or a National Standards Laboratory. A2LA accredited certification provides the highest achievable level of quality assurance, documentation, and accuracy. Conductivity Standards are produced in an A2LA accredited ISO/IEC 17025:2005 (1750.01) calibration laboratory. Additional certifications include ISO 31 (certificate content) and ISO 35 (statistical analysis), plus ISO 9001 (certified quality manufacturer).

NIST Traceable Calibration
Specifications
  • Conductivity Value (µS)1000
  • Conductivity Value (mS)1
  • TDS Value (ppm)666
  • Volume (mL)500
  • Volume (oz)16
  • DescriptionCERTIFIED CRM Conductivity and TDS Standard, Individually-Tested, 1000 µS; 500 mL
Reviews
Q&A