Environmental Express Ion Selective Electrode (ISE), Double-Junction, Glass; Sodium
Representative image only.
Environmental Express Ion Selective Electrode (ISE), Double-Junction, Glass; Sodium
Environmental Express | Item: 3580243
A wide variety of ion selective electrodes are availabe to suit a wide array of applications
₹96,500.00INR / Each
₹96,500.00INR / Each
Buy It Together
Current Item
Environmental Express Ion Selective Electrode (ISE), Double-Junction, Glass; Sodium
Environmental Express Ion Selective Electrode (ISE), Double-Junction, Glass; Sodium
3580243
₹96,500.00INR / Each
+
Cole-Parmer Copper (Cu+2) ISE Double junction solution kits
2750265
₹22,300.00INR / Each
+
Environmental Express EC 2000 Benchtop Meter Kit
3541960
₹185,000.00INR / Each
+
Cole-Parmer Replaceable Membrane ISE Probe; Ammonium
2750500
₹64,600.00INR / Each
+ + + + =
Total Price 788900.00 ADD 8 ITEMS TO CART
Product Description
Key Features
  • Epoxy-body types are either single- or double-junction
  • Glass-body type is double junction
  • 1 year warranty
More About this Item
Select from four electrode types: membrane, solid-state, gas sensing, and glass bulb; typical response time is 20 to 30 seconds.
1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Ion-Selective ElectrodeSodium
  • ConnectorBNC
  • DescriptionIon Selective Electrode (ISE), Double-Juntion, Glass; Sodium