Environmental Express Ion Selective Electrode (ISE), Double-Junction, Epoxy; Silver/Sulfide
Representative image only.
Environmental Express Ion Selective Electrode (ISE), Double-Junction, Epoxy; Silver/Sulfide
Environmental Express | Item: 3581240
A wide variety of ion selective electrodes are availabe to suit a wide array of applications
₹52,200.00INR / Each
₹52,200.00INR / Each
Buy It Together
Current Item
Environmental Express Ion Selective Electrode (ISE), Double-Junction, Epoxy; Silver/Sulfide
Environmental Express Ion Selective Electrode (ISE), Double-Junction, Epoxy; Silver/Sulfide
3581240
₹52,200.00INR / Each
+
Cole-Parmer Copper (Cu+2) ISE Double junction solution kits
2750265
₹22,300.00INR / Each
+
Cole-Parmer Replaceable Membrane ISE Probe; Ammonium
2750500
₹64,600.00INR / Each
+ + +
Environmental Express EC 2000 Benchtop Meter Kit
3541960
₹185,000.00INR / Each
+ + =
Total Price 744600.00 ADD 8 ITEMS TO CART
Product Description
Key Features
  • Epoxy-body types are either single- or double-junction
  • Glass-body type is double junction
  • 1 year warranty
More About this Item
Select from four electrode types: membrane, solid-state, gas sensing, and glass bulb; typical response time is 20 to 30 seconds.
1 Year Warranty
Specifications
  • Electrode TypeDouble junction, solid state
  • Electrode Body MaterialEpoxy
  • Ion-Selective ElectrodeSilver/Sulfide
  • ConnectorBNC
  • DescriptionIon Selective Electrode (ISE), Double-Juntion, Epoxy; Silver/Sulfide