Environmental Express HotBlock® TKN180 System Controller; Touchscreen
Representative image only.
Environmental Express HotBlock® TKN180 System Controller; Touchscreen
Environmental Express | Item: 3520569
Components to build your TKN system
₹1,536,000.00INR / Each
₹1,536,000.00INR / Each
Buy It Together
Current Item
Environmental Express HotBlock® TKN180 System Controller; Touchscreen
Environmental Express HotBlock® TKN180 System Controller; Touchscreen
3520569
₹1,536,000.00INR / Each
+ =
Total Price 3191000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • HotBlock®TKN block heaters available in 28- and 54-tube configurations
  • Controllers available as Simple or Advanced-Touchscreen units
  • 1 year warranty
More About this Item
Environmental Express HotBlock® TKN System provides an efficient method of digesting and storing water, wastewater, soil and sludge samples for Total Kjeldahl Nitrogen (TKN) analysis, total phosphorous (TP or TKP); total metals digestions, or other digestion methods requiring temperature up to 450°C. These innovative digestion systems allow samples to be digested in a corrosion-free environment. In addition, samples are handled in a small area with minimal radiant heat loss.
Touch-screen controller features: Status screen-displays parameters and status of each block currently running. Method Configuration Screen-used for programming and saving proceedures with user-defined parameters of ramp, set point temperature, soak time, and pause. Timer Screen- provides an automatic count down timer. Pause feature allows each block to be manually or programmed pause step during the digestion proceedure. Dimensions: 9 1/4" x 8 3/4".
1 Year Warranty
Specifications
  • Control TypeTouchscreen digital
  • Combination Block -Tube / Vial Size AcceptedUse with either 35205-66 or 35205-67 HotBlock®TKN block heaters
  • DescriptionEnvironmental Express HotBlock® TKN180 System Controller; Touchscreen