Environmental Express HotBlock® TKN100 System Controller; Simple
Representative image only.
Environmental Express HotBlock® TKN100 System Controller; Simple
Environmental Express | Item: 3520568
Components to build your TKN system
₹459,000.00INR / Each
₹459,000.00INR / Each
Buy It Together
Current Item
Environmental Express HotBlock® TKN100 System Controller; Simple
Environmental Express HotBlock® TKN100 System Controller; Simple
3520568
₹459,000.00INR / Each
+ =
Total Price 2114000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • HotBlock®TKN block heaters available in 28- and 54-tube configurations
  • Controllers available as Simple or Advanced-Touchscreen units
  • 1 year warranty
More About this Item
Environmental Express HotBlock® TKN System provides an efficient method of digesting and storing water, wastewater, soil and sludge samples for Total Kjeldahl Nitrogen (TKN) analysis, total phosphorous (TP or TKP); total metals digestions, or other digestion methods requiring temperature up to 450°C. These innovative digestion systems allow samples to be digested in a corrosion-free environment. In addition, samples are handled in a small area with minimal radiant heat loss.
Program other methods or variations of the TNK method into the TKN basic controller.
1 Year Warranty
Specifications
  • Control TypeDigital
  • Combination Block -Tube / Vial Size AcceptedUse with either 35205-66 or 35205-67 HotBlock®TKN block heaters
  • DescriptionEnvironmental Express HotBlock® TKN100 System Controller; Simple