Environmental Express Grit Pot, Aluminum Cyclone; 25/Pk
Representative image only.
Environmental Express Grit Pot, Aluminum Cyclone; 25/Pk
Environmental Express | Item: 6006194
Help aluminum cyclones work at its best
₹2,600.00INR / Pkg of 25
₹2,600.00INR / Pkg of 25
Product Description
Key Features
  • Fits on the cyclone to collect the larger particles
  • Works with both 25- and 37-mm cyclones
Specifications
  • Compatible With25- and 37-mm aluminum cyclones
  • DescriptionZefon Grit Pot, Aluminum Cyclone; 25/PK