Environmental Express G7004C Flow-Control Valve for QuikVap Evaporating System
Representative image only.
Environmental Express G7004C Flow-Control Valve for QuikVap Evaporating System
Environmental Express | Item: 5310001
Replacement parts keep your system running
₹12,600.00INR / Each
₹12,600.00INR / Each
Product Description
Key Features
  • Evaporate n-hexane quickly and effectively for oil and grease testing
  • Model number G7004C
More About this Item
Versatile system evaporates n-hexane quickly and effectively with adjustable temperature control, airflow control, and vessel configuration. Use digital controller to accurately set and maintain temperature throughout the evaporation process.
Replacement part for the QuikVap Evaporating System (G7000). The flow-control valve regulates the amount of air or nitrogen moving through the curved needles. The flow control valve connects to the Hose Barb Adaptor (G5000).
Specifications
  • Compatible WithQuikVap Evaporating System
  • DescriptionFlow-Control Valve for QuikVap Evaporating System