Environmental Express FilterMate™ Replacement Plungers, 50 mL; 500/Pk
Representative image only.
Environmental Express FilterMate™ Replacement Plungers, 50 mL; 500/Pk
Environmental Express | Item: 3520207
No need for a brand new FilterMate
₹36,400.00INR / Pkg of 500
₹36,400.00INR / Pkg of 500
Product Description
Key Features
  • Plunger or sample tube available
  • Works with 50 mL sample tube and FilterMate
  • 3 month warranty
More About this Item
The machined HDPE holder adds stability when performing FilterMate filtrations. The 50 mL sample tube is placed into the center hole of the FilterMate holder prior to filtration. The holder secures the digestion tube as the FilterMate is depressed through the liquid sample.
Replacement plungers, for 50 mL cup. Pack of 500.
3 Month Warranty
Specifications
  • MaterialPolypropylene (PP)
  • Compatible With50 mL digestion tubes
  • DescriptionReplacement Plungers, 50 mL; 500/Pk