Environmental Express E081111C Cubitainer, HDPE, 1 Liter; 128/Cs
Representative image only.
Environmental Express E081111C Cubitainer, HDPE, 1 Liter; 128/Cs
| Item: 3520875
Collapsible containers nest for efficient storage
₹111,000.00INR / Case of 128
₹111,000.00INR / Case of 128
Product Description
Includes
polypropylene caps with foamed PE liner.
Key Features
  • Meets Level 3 classification for contaminant-free sample containers
  • Just unscrew cap and fill to expand to total volume
  • Model number E081111C
More About this Item
Cubitainer® containers meet EPA recommended guidelines for contamination-free sample containers, but do not ship with a certificate of analysis.
Specifications
  • Capacity (mL)1000
  • Capacity (L)1
  • Capacity (oz)32
  • Capacity (Gal)0.26
  • MaterialHigh Density Polyethyene (HDPE)
  • Bottle ColorNatural
  • AutoclavableNo
  • DescriptionCubitainer, HDPE, 1 Liter; 128/Cs