Environmental Express E081101C Cubitainer, HDPE, 4 Liter; 160/Cs
Representative image only.
Environmental Express E081101C Cubitainer, HDPE, 4 Liter; 160/Cs
| Item: 3520868
Collapsible containers nest for efficient storage
₹152,000.00INR / Case of 160
₹152,000.00INR / Case of 160
Product Description
Includes
polypropylene caps with foamed PE liner.
Key Features
  • Meets Level 3 classification for contaminant-free sample containers
  • Just unscrew cap and fill to expand to total volume
  • Model number E081101C
More About this Item
Cubitainer® containers meet EPA recommended guidelines for contamination-free sample containers, but do not ship with a certificate of analysis.
Specifications
  • Capacity (mL)4000
  • Capacity (L)4
  • Capacity (oz)128
  • Capacity (Gal)1
  • MaterialHigh Density Polyethyene (HDPE)
  • Bottle ColorNatural
  • AutoclavableNo
  • DescriptionCubitainer, HDPE, 4 Liter; 160/Cs