Environmental Express D1055 BOD Insert for Thermo® Orion BOD Analyzer, 12-Place
Representative image only.
Environmental Express D1055 BOD Insert for Thermo® Orion BOD Analyzer, 12-Place
Environmental Express | Item: 0800276
Racks that fit the Thermo® BOD analyzer or Skalar® BOD analyzer
₹11,700.00INR / Each
₹11,700.00INR / Each
Product Description
Key Features
  • Works with Environmental Express® Disposable BOD Bottles (D1001) Not suitable for glass bottles
  • Model number D1055
More About this Item
D1055 rack has 12-place inserts that fit into Thermo® Orion analyzer racks to center disposable BOD bottles. Inserts are made from 1/2" high density polyethylene.
Specifications
  • MaterialPolyethylene(PE)
  • ColorBlack
  • DescriptionBOD Insert for Thermo® Orion BOD Analyzer, 12-Place