Environmental Express Combination pH electrode, Glass, Refillable, Double-Junction, BNC
Representative image only.
Environmental Express Combination pH electrode, Glass, Refillable, Double-Junction, BNC
Environmental Express | Item: 5900134
Faster response and increased stability across a pH range of 0 to 14
₹34,200.00INR / Each
₹34,200.00INR / Each
Buy It Together
Current Item
Environmental Express Combination pH electrode, Glass, Refillable, Double-Junction, BNC
Environmental Express Combination pH electrode, Glass, Refillable, Double-Junction, BNC
5900134
₹34,200.00INR / Each
+ =
Total Price 45400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
  • 1 year warranty
More About this Item
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.
1 Year Warranty
Specifications
  • ApplicationsResearch, TRIS, sulfates, proteins
  • Electrode TypeDouble junction
  • Electrode Body MaterialGlass
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)100
  • ConnectorBNC
  • DescriptionCombination pH electrode, Glass, Refillable, Double-Junction, BNC