Environmental Express Cole Parmer PC200S pH/Conductivity Meter Kit; Cell, Tris pH Electrode, Test Bench

Representative image only.

Environmental Express Cole Parmer PC200S pH/Conductivity Meter Kit; Cell, Tris pH Electrode, Test Bench

Environmental Express | Item: 5925025

Options for every application

₹190,000.00INR / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹190,000.00INR / Each

Buy It Together

Current Item

Environmental Express Cole Parmer PC200S pH/Conductivity Meter Kit; Cell, Tris pH Electrode, Test Bench
Environmental Express Cole Parmer PC200S pH/Conductivity Meter Kit; Cell, Tris pH Electrode, Test Bench
5925025

₹190,000.00INR / Each
+ + +
Environmental Express Oakton PH 550 Benchtop pH Meter Kit
3541930

₹75,500.00INR / Each
+ +
Environmental Express 1500 pH Benchtop Meter Kit
3541940

₹87,400.00INR / Each
+ +
Cole-Parmer 100-series Replacement pH/Temperature Electrode
5920006

₹22,100.00INR / Each
+ =
Total Price 643100.00 ADD 9 ITEMS TO CART

Customers Also Bought

Product Description

Includes

pH electrode, conductivity electrode, test bench, and power cord.

Key Features

  • Choice of electrodes—combination 3-in-1 with ATC or Tris-compatible
  • Keep your workspace tidy with the test bench ("S" models only)

More About this Item

The P200 series of benchtop meters provides accurate, reliable results with the benefit of convenient accessories to make your lab run smoothly. Large color TFT display is easily visible, and features a stability icon to alert you when your readings are ready to be recorded and an electrode health icon indicating when you need to refill or replace your electrode.

The test bench features a magnetic stirrer (0 to 3000 rpm) and flexible-arm electrode holder, with dedicated spaces for calibration standards or sample cups and a stir bar. The intelligent stirrer has memory for repeatable results and must be used with meter to power and control stirrer functions.

Not for sale in Canada!

3 Year Warranty
Specifications
  • Meter TypeBenchtop with built-in stirrer, 0 to 3000 rpm
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01, 0.1
  • pH Accuracy±0.01
  • Buffer RecognitionpH: USA, NIST, user defined; Conductiviyt: 84, 1413 µS; 12.88, 111.8 mS; one user defined
  • CalibrationpH: 1 to 3 points; Conductivity: 1 to 4 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Accuracy±0.01% full-scale
  • Max Conductivity200 mS
  • Conductivity Resolution0.01, 0.1 (µS/cm or mS/cm)
  • Conductivity Accuracy±1% full-scale
  • CellIncluded
  • Min TDS0.0 mg/L
  • Max TDS100.0 g/L
  • TDS Resolution0.1
  • TDS Accuracy±1% full-scale
  • Min Temperature (° C)0
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature Resolution0.1°F/C
  • Temperature Accuracy±0.36°F (0.2°C)
  • Temperature compensationYes, 32 to 212°F (0 to 100°C), automatic or manual
  • Power (VAC)110 - 120
  • Power (Hz)60
  • DescriptionPC200S pH/Conductivity Meter Kit; Cell, Tris pH Electrode, Test Bench
Reviews
Q&A