Environmental Express Cole Parmer PC200-02 pH/Conductivity Meter Kit; Cell, 3-in-1 pH Electrode, Holder

Representative image only.

Environmental Express Cole Parmer PC200-02 pH/Conductivity Meter Kit; Cell, 3-in-1 pH Electrode, Holder

Environmental Express | Item: 5925015

Options for every application

₹165,000.00INR / Each

₹165,000.00INR / Each

Buy It Together

Current Item

Environmental Express Cole Parmer PC200-02 pH/Conductivity Meter Kit; Cell, 3-in-1 pH Electrode, Holder
Environmental Express Cole Parmer PC200-02 pH/Conductivity Meter Kit; Cell, 3-in-1 pH Electrode, Holder
5925015

₹165,000.00INR / Each
+ + + + +
Environmental Express 1500 pH Benchtop Meter Kit
3541940

₹87,400.00INR / Each
+ +
Cole-Parmer 100-series Replacement pH/Temperature Electrode
5920006

₹22,100.00INR / Each
+ =
Total Price 637800.00 ADD 9 ITEMS TO CART

Customers Also Bought

Product Description

Includes

conductivity cell, pH electrode, flexible-arm electrode holder, and power cord.

Key Features

  • Choice of electrodes—combination 3-in-1 with ATC or Tris-compatible
  • Flexible-arm electrode holder lets you perform other tasks while waiting for readings to stabilize

More About this Item

The P200 series of benchtop meters provides accurate, reliable results with the benefit of convenient accessories to make your lab run smoothly. Large color TFT display is easily visible, and features a stability icon to alert you when your readings are ready to be recorded and an electrode health icon indicating when you need to refill or replace your electrode.

The pH/conductivity meters ("PC" models) display both pH and conductivity simultaneously.

3 Year Warranty
Specifications
  • Meter TypeBenchtop
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01, 0.1
  • pH Accuracy±0.01
  • Buffer RecognitionpH: USA, NIST, user defined; Conductiviyt: 84, 1413 µS; 12.88, 111.8 mS; one user defined
  • CalibrationpH: 1 to 3 points; Conductivity: 1 to 4 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Accuracy±0.01% full-scale
  • Max Conductivity200 mS
  • Conductivity Resolution0.01, 0.1 (µS/cm or mS/cm)
  • Conductivity Accuracy±1% full-scale
  • CellIncluded
  • Min TDS0.0 mg/L
  • Max TDS100.0 g/L
  • TDS Resolution0.1
  • TDS Accuracy±1% full-scale
  • Min Temperature (° C)0
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Temperature Resolution0.1°F/C
  • Temperature Accuracy±0.36°F (0.2°C)
  • Temperature compensationYes, 32 to 212°F (0 to 100°C), automatic or manual
  • Power (VAC)110 - 120
  • Power (Hz)60
  • DescriptionPC200-02 pH/Conductivity Meter Kit; Cell, 3-in-1 pH Electrode, Holder
Reviews
Q&A