Environmental Express COD Tubes, 0 to 150ppm, EPA; 100/Pk

Representative image only.

Environmental Express COD Tubes, 0 to 150ppm, EPA; 100/Pk

Environmental Express | Item: 3520002

Obtain consistent and accurate results while saving money

Call For Price

Call For Price

Buy It Together

Current Item

Environmental Express COD Tubes, 0 to 150ppm, EPA; 100/Pk
Environmental Express COD Tubes, 0 to 150ppm, EPA; 100/Pk
3520002

Call For Price
+
Environmental Express Model 200 COD Colorimeter
9956184

₹176,000.00INR / Each
=
Total Price 176000.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Certificate of Analysis and SDS.

Key Features

  • Pre-filled reagent vials for either EPA-approved or mercury-free COD analysis
  • Quality analyzed to provide you hassle-free consistent results
  • Don''t change your instrument or method—vials work with Hach, Orbeco, Chemetrics, and Thermo COD systems!
  • Comparable Hach number 21258-15/25

More About this Item

Pre-filled with COD reagents, these digestion vials come in either EPA-approved or mercury-free versions for low-, standard-, and ultra-high-range analysis. Each lot is tested to ensure superior quality from the cap and vial to the chemical contents. Glass vials are 16 mm x 100 mm to work with your existing COD reactor and spectrophotometer. Simply add 2 mL of your sample, heat in your digestor, and read in your spectrometer.

Although mercury-free vials are not approved for EPA Methods, they may be used for non-EPA COD testing when chlorine interferrence is not a factor. Ultra-high-range reagent vials have also not approved for EPA testing methods. COD digestion vials may contain acid, mercury, and silver and must be disposed of properly.

Warning: Contains mercury (Hg). Do not dispose of in household waste. Dispose of in accordance with applicable local, national, or international regulations.

EPA United States Environmental protection agency
Specifications
  • TestChemical oxygen demand (COD)
  • Measurement Points0 to 150 ppm
  • Reagent PlatformEPA Approved
  • Tests Per Kit100
  • DescriptionTubes, 0 to 150ppm, EPA; 100/Pk
Reviews
Q&A