Environmental Express COD Digestion Vials w/Hg, 0-1500mg/L, 98pk
Representative image only.
Environmental Express COD Digestion Vials w/Hg, 0-1500mg/L, 98pk
Environmental Express | Item: 1596999
Obtain consistent and accurate results while saving money
₹40,700.00INR / Pkg of 98
₹40,700.00INR / Pkg of 98
Buy It Together
Current Item
Environmental Express COD Digestion Vials w/Hg, 0-1500mg/L, 98pk
Environmental Express COD Digestion Vials w/Hg, 0-1500mg/L, 98pk
1596999
₹40,700.00INR / Pkg of 98
+
Environmental Express Model 200 COD Colorimeter
9956184
₹176,000.00INR / Each
=
Total Price 216700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Certificate of Analysis and SDS.
Key Features
  • Pre-filled reagent vials for either EPA-approved or mercury-free COD analysis
  • Quality analyzed to provide you hassle-free consistent results
  • Don''t change your instrument or method—vials work with Hach, Orbeco, Chemetrics, and Thermo COD systems!
More About this Item
Pre-filled with COD reagents, these digestion vials come in either EPA-approved or mercury-free versions for low-, standard-, and ultra-high-range analysis. Each lot is tested to ensure superior quality from the cap and vial to the chemical contents. Glass vials are 16 mm x 100 mm to work with your existing COD reactor and spectrophotometer. Simply add 2 mL of your sample, heat in your digestor, and read in your spectrometer.
Although mercury-free vials are not approved for EPA Methods, they may be used for non-EPA COD testing when chlorine interferrence is not a factor. Ultra-high-range reagent vials have also not approved for EPA testing methods. COD digestion vials may contain acid, mercury, and silver and must be disposed of properly.