Environmental Express Buffer Solution, pH 10.01; 1000 mL

Representative image only.

Environmental Express Buffer Solution, pH 10.01; 1000 mL

Environmental Express | Item: 0594262

Calibrate pH instruments to ensure accuracy

₹2,900.00INR / Each

₹2,900.00INR / Each
Product Description

Key Features

  • Lab-grade buffers available in multiple volumes to reduce waste and support various levels of throughput
  • Color coded and/or clearly labeled for easy identification in busy labs
  • Unopened, shelf life is 2 years
  • USA buffer value of pH 10.01

More About this Item

pH 10.01 is a common low-range pH meter calibration point. All Environmental Express pH buffer solutions are manufactured under ISO 9000 quality guidelines and traceable to NIST standard (for meters with 0.01 resolution). A pH/temperature table is printed on the label for accurate calibrations.

pH 10.01 buffer solution is colored red for visual reference. It is sold in individual 1000-mL (1-L) bottles. If unopened, shelf life of pH 10.01 is 2 years.

Store buffers away from direct sunlight.

Specifications
  • Buffer Value10.01
  • Volume (mL)1000
  • ColorBlue
  • DescriptionBuffer Solution, pH 10.01; 1000 mL
Documents & Certificates

Certificate of Analysis

Enter lot number
or click to view a Sample C of A
Reviews
Q&A