Environmental Express Assembled Filter Funnel, PVDF, 0.45 um, 250/pk
Representative image only.
Environmental Express Assembled Filter Funnel, PVDF, 0.45 um, 250/pk
Environmental Express | Item: 3520037
Prevent sample-to-sample contamination to ensure accurate alpha particle spectra
₹185,000.00INR / Pkg of 1
₹185,000.00INR / Pkg of 1
Product Description
Key Features
  • Polypropylene construction withstands exposure to acids and bases
  • Choose convenient preassembled filter units or separate funnels and filters to accommodate any protocol
More About this Item
Total funnel capacity is 68 mL and filter diameter is 25 mm. Funnel stem fits a standard rubber stopper hole. Easily remove the filter to analyze filtrate.
Assembled filter funnels with filters include a top cap, filter support and filter.
Specifications
  • Membrane MaterialPolyvinylidene fluoride (PVDF)
  • Pore Size (µm)0.45
  • Membrane Diameter (mm)25
  • Sample Volume (mL)68
  • DescriptionAssembled Filter Funnel, PVDF, 0.45 um, 250/pk