Environmental Express All-in-One pH/ATC Probe, Refillable/SJ/Epoxy; for Series
Representative image only.
Environmental Express All-in-One pH/ATC Probe, Refillable/SJ/Epoxy; for Series
Environmental Express | Item: 5900274
₹23,300.00INR / Each
₹23,300.00INR / Each
Buy It Together
Current Item
Environmental Express All-in-One pH/ATC Probe, Refillable/SJ/Epoxy; for Series
Environmental Express All-in-One pH/ATC Probe, Refillable/SJ/Epoxy; for Series
5900274
₹23,300.00INR / Each
+ =
Total Price 29200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Use to replace pH/ATC electrodes for discontinued Oakton pH 10, 100, 200, 500, 1000, 2000, 2500 series meters.
1 Year Warranty
Specifications
  • ApplicationsRefillable
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Temperature CompensationYes
  • ATC ConnectorAudio plug, 3.5 mm
  • ConnectorBNC
  • DescriptionAll-in-One pH/ATC Probe, Refillable/SJ/Epoxy; for Series