Environmental Express 5 mL Vial with Filtering Cap for Dionex® AS-40 or AS-DV Autosamplers; 250/PK
Representative image only.
Environmental Express 5 mL Vial with Filtering Cap for Dionex® AS-40 or AS-DV Autosamplers; 250/PK
Environmental Express | Item: 5005454
Vials constructed to fit Dionex® Autosamplers or Metrohm IC instrumentation
₹19,700.00INR / Pkg of 250
₹19,700.00INR / Pkg of 250
Product Description
Key Features
  • These polypropylene straight-sided vials fit directly into the sample cassettes on the AS-40 or AS-DV autosampler
  • Filter caps are molded of LDPE and have a porous filter plug
  • Superior filter cap design aligns perfectly into top of 5 mL vial so the cap is always straight
  • Model number K1250
Specifications
  • Volume (mL)5
  • Tube MaterialPolypropylene
  • SterileNo
  • Description5 mL Vial with Filtering Cap for Dionex® AS-40 or AS-DV Autosamplers; 250/PK