Environmental Express 1.5 mL Vial and Cap for Dionex® AS-50 and AS-AP Autosamplers; 100/PK
Representative image only.
Environmental Express 1.5 mL Vial and Cap for Dionex® AS-50 and AS-AP Autosamplers; 100/PK
Environmental Express | Item: 5005467
Vials constructed to fit Dionex® Autosamplers or Metrohm IC instrumentation
₹14,800.00INR / Pkg of 100
₹14,800.00INR / Pkg of 100
Product Description
Key Features
  • Polypropylene vial with septum cap for Dionex AS-50 and AS-AP Autosampler
  • The septum is slit for easy penetration
  • The septum is a laminate of blue PTFE on the bottom and white silcone on the top
  • Model number K4350
Specifications
  • Volume (mL)1.5
  • Tube MaterialPolypropylene
  • SterileNo
  • Description1.5 mL Vial and Cap for Dionex® AS-50 and AS-AP Autosamplers; 100/PK