Environmental Devices HD-1100 Environemental Devices Haz-Dust Real-Time Dust Monitor
Representative image only.
Environmental Devices HD-1100 Environemental Devices Haz-Dust Real-Time Dust Monitor
Environmental Devices | Mfr: HD-1100| Item: 8655300
Provides instant airborne particulate concentrations in accordance with NIOSH method #0600
Call For Price
Call For Price
Product Description
Key Features
  • Measures concentration levels from 0.01 up to 200 mg/m3
  • Ideal for industrial hygiene, indoor air quality, and haz-mat monitoring
  • 1 year warranty
More About this Item
Affordable, hand-held survey monitor is lightweight and easy to operate. Simply turn monitor on and immediately view dust concentrations on digital display. Sensing range is easily selected from 0.1 to 200 or 0.01 to 20 mg/m3 at the flip of a switch . Haz-Dust monitor is appropriate for a variety of aerosols and dusts including: lead, silica, coal, wood, dry chemicals, welding fumes, smoke, paint spray, and cadmium.
Rugged, well-constructed ABS plastic case protects monitor electronics from accidental drops. Unique design provides protection against RFI/EMI waves.
What's included: rechargeable battery with 110/230 VAC, 50/60 Hz charger and carrying case.
1 Year Warranty
Specifications
  • Particle Size (micron)0.01 to 50
  • Display3-1/2-digit LCD
  • PowerRechargeable NiMH battery with 110/230 VAC, 50/60 Hz charger
  • Battery Life>8 hours
  • DescriptionEnvironemental Devices Haz-Dust Real-Time Dust Monitor