Enduro E0168 Standard casting set for model Gel XL Electrophoresis System
Representative image only.
Enduro E0168 Standard casting set for model Gel XL Electrophoresis System
Enduro | Mfr: E0168| Item: 1731421
Optimize performance of your gel horizontal electrophoresis system
₹52,200.00INR / Pkg of 1
₹52,200.00INR / Pkg of 1
Product Description
Key Features
  • Keep your electrophoresis system running properly with these accessories
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Enduro gel XL horizontal electrophoresis system.
Includes one large gel tray, two small gel trays, four large combs, and casting stand.
1 Year Warranty
Specifications
  • DescriptionStandard casting set for model Gel XL Electrophoresis System