Enduro E0165 Small comb set, 5/8 teeth, 2-pack for Gel XL Electrophoresis System
Representative image only.
Enduro E0165 Small comb set, 5/8 teeth, 2-pack for Gel XL Electrophoresis System
Enduro | Mfr: E0165| Item: 1731427
Optimize performance of your gel horizontal electrophoresis system
₹10,100.00INR / Pkg of 2
₹10,100.00INR / Pkg of 2
Product Description
Key Features
  • Keep your electrophoresis system running properly with these accessories
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Enduro gel XL horizontal electrophoresis system.
The micro combs are required in order to run up to four micro gels (6 x 6 cm) in the Enduro Gel XL Electrophoresis system (17314-11, -13,-15). Includes 5/8 teeth, which are reversible. Set of 2.
1 Year Warranty
Specifications
  • DescriptionSmall comb set, 5/8 teeth, 2-pack for Gel XL Electrophoresis System