Encon 5058204 Spherical Goggles, Clear Antifogging Lens
Representative image only.
Encon 5058204 Spherical Goggles, Clear Antifogging Lens
Encon | Mfr: 05058204| Item: 0166214
Special lens eliminates distortion
₹2,200.00INR / Each
₹2,200.00INR / Each
Product Description
Key Features
  • Air vents repel splashes and contaminants and ensure continuous airflow
  • Flexible frames and adjustable neoprene headbands for a snug fit
More About this Item
Compliance: ANSI Z87.1-2003, CSA Z94.3-92, SEI certified
Canadian Standards Association
Specifications
  • Frame ColorClear
  • Lens ColorClear
  • Lens CoatingAntifog
  • ComplianceANSI Z87.1-2003; CSA Z94.3-92; SEI-certified
  • DescriptionSpherical Goggles, Clear Antifogging Lens