EMD Millipore UFC805024 Amicon Ultra Centrifugal Filters, 4 mL, 50K MWCO; pack of 24

Representative image only.

EMD Millipore UFC805024 Amicon Ultra Centrifugal Filters, 4 mL, 50K MWCO; pack of 24

EMD Millipore | Mfr: UFC805024| Item: 2996976

Low-binding regenerated cellulose membrane, fast processing, and >90% recoveries

₹82,500.00INR / Each

₹82,500.00INR / Each
Product Description

Includes

24 units pre-assembled with centrifuge tubes and caps.

Key Features

  • Unique vertical housing
  • Designed with 4.0-mL volume and 50,000 Nominal molecular weight limit (NMWL)
  • 1 year warranty

More About this Item

These units are premier tools for protein concentration and desalting. Typical applications include purifying macromolecular components from tissue culture extracts or cell lysates and concentrating biological samples.

1 Year Warranty
Specifications
  • Nominal Molecular Weight Limit (Mwl) (Dalton)50000
  • Tube Size (mL)4
  • DescriptionAmicon Ultra Centrifugal Filters, 4 mL 50K 24PK
  • Warranty1 year
Reviews
Q&A