EMD Millipore UFC505024 Amicon Ultra Centrifugal Filters, 0.5 mL, 50K MWCO; pack of 24

Representative image only.

EMD Millipore UFC505024 Amicon Ultra Centrifugal Filters, 0.5 mL, 50K MWCO; pack of 24

EMD Millipore | Mfr: UFC505024| Item: 2996826

Low-binding regenerated cellulose membrane, fast processing, and >90% recoveries

₹49,500.00INR / Pkg of 24

₹49,500.00INR / Pkg of 24
Product Description

Includes

24 units pre-assembled with centrifuge tubes and caps.

Key Features

  • Unique vertical housing
  • Designed with 0.5-mL volume and 50,000 Nominal molecular weight limit (NMWL)

More About this Item

These units are premier tools for protein concentration and desalting. Typical applications include purifying macromolecular components from tissue culture extracts or cell lysates and concentrating biological samples.

Specifications
  • Max Retention Volume (µL)20
  • Max Sample Volume (mL)0.5
  • Min Retention Volume (µL)15
  • Nominal Molecular Weight Limit (Mwl) (Dalton)50000
  • Tube Size (mL)0.5
  • DescriptionAmicon Ultra Centrifugal Filters, 0.5 mL, 50K MWCO; pack of 24
Reviews
Q&A