EMD Millipore UFC500324 Amicon Ultra Centrifugal Filters, 0.5 mL, 3K NMWL; pack of 24

Representative image only.

EMD Millipore UFC500324 Amicon Ultra Centrifugal Filters, 0.5 mL, 3K NMWL; pack of 24

EMD Millipore | Mfr: UFC500324| Item: 2996820

Low-binding regenerated cellulose membrane, fast processing, and >90% recoveries

₹72,100.00INR / Pkg of 24

₹72,100.00INR / Pkg of 24
Product Description

Includes

24 units pre-assembled with centrifuge tubes and caps.

Key Features

  • Unique vertical housing
  • Designed with 0.5-mL volume and 3,000 Nominal molecular weight limit (NMWL)

More About this Item

These units are premier tools for protein concentration and desalting. Typical applications include purifying macromolecular components from tissue culture extracts or cell lysates and concentrating biological samples.

Specifications
  • Max Retention Volume (µL)20
  • Nominal Molecular Weight Limit (Mwl) (Dalton)3000
  • Max Sample Volume (mL)0.5
  • Min Retention Volume (µL)15
  • Tube Size (mL)0.5
  • DescriptionAmicon Ultra Centrifugal Filters, 0.5 mL, 3K NMWL; pack of 24
Reviews
Q&A