EMD Millipore PLTK04310 Ultrafiltration Disc Membranes, 30k 43mm 10pk

Representative image only.

EMD Millipore PLTK04310 Ultrafiltration Disc Membranes, 30k 43mm 10pk

EMD Millipore | Mfr: PLTK04310| Item: 2994926

Ideal for stirred cells and micropartition system

Call For Price

Call For Price
Product Description

Key Features

  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
  • 1 year warranty

More About this Item

These high-recovery hydrophilic membranes are made of regenerated cellulose. Use when concentrating or desalting extremely dilute solutions or whenever your sample is hydrophobic.

1 Year Warranty
Specifications
  • Nominal Molecular Weight Limit (Mwl) (Dalton)30000
  • Diameter (mm)44.5
  • Diameter (cm)4.45
  • DescriptionUltrafiltration Disc Membranes, 30k 43mm 10pk
  • Warranty1 year
Reviews
Q&A