EMD Millipore HYCON® 941605 Blank Agar Strips; 50/Box
Representative image only.
EMD Millipore HYCON® 941605 Blank Agar Strips; 50/Box
EMD Millipore | Mfr: 1441070050| Item: 8650960
Choose from strips with or without media
₹53,300.00INR / Box of 50
₹53,300.00INR / Box of 50
Product Description
Key Features
  • Customize by adding your own media
More About this Item
Compatible with the Biotest Hycon RCS® Air Sampler 86509-00, the sampler operates on impaction principle, drawing airborne microorganisms onto an agar media strip for later incubation and analysis.
The Blank Strip Kit consists of 50 empty, sealed strips manufactured in a controlled environment. It is intended for applications where ready-to-use HYCON® agar strips do not meet the user’s requirements. Blank strips are coated with self-made, specialized agar media.
Sampler sold separately.
Specifications
  • Compatible WithBiotest Hycon RCS Air Sampler
  • DescriptionBlank Agar Strips; 50/Box