Eldon James Hose Barb Fitting, Y Union, Stainless Steel, 3/8" ID; 1/Each
Representative image only.
Eldon James Hose Barb Fitting, Y Union, Stainless Steel, 3/8" ID; 1/Each
Eldon James | Mfr: Y0-6SS| Item: 5012800
Excellent for high temperature and harsh physical environments
₹8,400.00INR / Each
₹8,400.00INR / Each
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
  • 3/8" hose barb
More About this Item
Easily combine or split fluid flow using Y unions. These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialStainless Steel
  • Sterilization MethodE-beam/gamma irradiation, EtO, or autoclave up to 134°C
  • Connection TypeHose Barb Union
  • Number Of Ports3
  • Barb Size / Tubing Id (Port 1) (in)3/8
  • Barb Size / Tubing Id (Port 2) (in)3/8
  • Barb Size / Tubing Id (Port 3) (in)3/8
  • Flow PatternY
  • ColorMetallic
  • Regulatory ApprovalsRoHS;Non-DEHP;Non-Phthalate
  • DescriptionHose Barb Fitting, Y Union, Stainless Steel, 3/8" ID; 1/Ea