Eldon James Hose Barb Fitting, Straight Union, Natural Kynar®, 5/16" ID; 10/PK
Representative image only.
Eldon James Hose Barb Fitting, Straight Union, Natural Kynar®, 5/16" ID; 10/PK
Eldon James | Mfr: C0-5NK7| Item: 5012506
Single barb design tightly “grips” tubing
₹4,200.00INR / Pkg of 10
₹4,200.00INR / Pkg of 10
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Biopharma compatible and USP 661 and BPOG compliant
  • 5/16" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose use applications. Easily connect two tubing with straight unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF
  • Connection TypeHose Barb Union
  • Number Of Ports2
  • Barb Size / Tubing Id (Port 1) (in)5/16
  • Barb Size / Tubing Id (Port 2) (in)5/16
  • Flow PatternStraight
  • ColorNatural
  • Min Temperature (° F)-31
  • Max Temperature (° F)275
  • Min Temperature (° C)-35
  • Max Temperature (° C)135
  • Regulatory ApprovalsNon-DEHP;Non-Phthalate;USP
  • DescriptionHose Barb Fitting, Straight Union, Natural Kynar®, 5/16" ID; 10/PK