Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 3/32" ID; 10/PK
Representative image only.
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 3/32" ID; 10/PK
Eldon James | Mfr: C0-1.5-200PP| Item: 5012476
Single barb design tightly “grips” tubing
₹1,300.00INR / Pkg of 10
₹1,300.00INR / Pkg of 10
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
  • 3/32" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose use applications. Easily connect two tubing with straight unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolypropylene
  • Number Of Ports2
  • Connection TypeHose Barb Union
  • Barb Size / Tubing Id (Port 1) (in)3/32
  • Barb Size / Tubing Id (Port 2) (in)3/32
  • Flow PatternStraight
  • ColorNatural
  • Min Temperature (° F)-9
  • Max Temperature (° F)150
  • Min Temperature (° C)-23
  • Max Temperature (° C)66
  • Regulatory ApprovalsRoHS;Non-DEHP;FDA;Non-Phthalate;ADCF
  • DescriptionHose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 3/32" ID; 10/PK